4-(Aminomethyl)benzoic acid, 99%
4-(Aminomethyl)benzoic acid, 99%
4-(Aminomethyl)benzoic acid, 99%
Thermo Scientific Chemicals

4-(Aminomethyl)benzoic acid, 99%

CAS: 56-91-7 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 232221000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS56-91-7
IUPAC Name4-(aminomethyl)benzoic acid
Molecular FormulaC8H9NO2
InChI KeyQCTBMLYLENLHLA-UHFFFAOYSA-N
SMILESNCC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.5 %
Water=<6 % (K.F.)
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only