Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Create a compact, space saving, reagent dispensing bank using chemical-resistant Thermo Scientific™ Nalgene™ Low-Profile HDPE Lowboys. The low-profile design of these high density polyethylene lowboys fits into tight spaces on shelves and in refrigerators and can stack three high. Chemically compatible with most aqueous and alcohol lab liquids, including reagents, distilled water and titrants. Convenient integral carry handle for safe, easy transport. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Capacity (English) | 4 gal. |
Closure Material | Polypropylene |
Capacity (Metric) | 15 L |
Material | HDPE |
Type | Low-Profile Carboy |
For Use With (Application) | Dispense most liquids, including reagents, distilled water and titrants. |
Autoclavable | Not Autoclavable |
Color | White |
Unit Size | Each |
Capacity (English) | 4 gal. |
Closure Material | Polypropylene |
Capacity (Metric) | 15 L |
Material | HDPE |
Type | Low-Profile Carboy |
For Use With (Application) | Dispense most liquids, including reagents, distilled water and titrants. |
Autoclavable | Not Autoclavable |
Color | White |
Unit Size | Case of 4 |
Capacity (English) | 2 gal. |
Closure Material | Polypropylene |
Capacity (Metric) | 8 L |
Material | HDPE |
Type | Low-Profile Carboy |
For Use With (Application) | Dispense most liquids, including reagents, distilled water and titrants. |
Autoclavable | Not Autoclavable |
Color | White |
Unit Size | Case of 4 |
Capacity (English) | 2 gal. |
Closure Material | Polypropylene |
Capacity (Metric) | 8 L |
Material | HDPE |
Type | Low-Profile Carboy |
For Use With (Application) | Dispense most liquids, including reagents, distilled water and titrants. |
Autoclavable | Not Autoclavable |
Color | White |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2323-0015PK also known as 2323-0015 | Each | 15 L | Price: 264.10 Your Price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2323-0015 | Case of 4 | 15 L | Price: 895.00 Your Price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2323-0008 | Case of 4 | 8 L | Price: 786.00 Your Price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2323-0008PK also known as 2323-0008 | Each | 8 L | Price: 232.00 Your Price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Includes:
Spigot and spigot closure and four acid-proof, pressure-sensitive labels
Note:
Nalgene rugged HDPE carboys are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE carboys and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene carboy products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE carboys.