Linalyl acetate, 95%, synthetic
Linalyl acetate, 95%, synthetic
Linalyl acetate, 95%, synthetic
Thermo Scientific Chemicals

Linalyl acetate, 95%, synthetic

CAS: 115-95-7 | C12H20O2 | 196.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 232305000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS115-95-7
IUPAC Name3,7-dimethylocta-1,6-dien-3-yl acetate
Molecular FormulaC12H20O2
InChI KeyUWKAYLJWKGQEPM-UHFFFAOYNA-N
SMILESCC(C)=CCCC(C)(OC(C)=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Density(25°C) 0.895 to 0.914 g/cm³
GC>=94 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only