See complete Detergent-compatible Bradford Assay Kit, Cat # 23246
Thermo Scientific™

Pierce™ Detergent Compatible Bradford Assay Reagent

Catalog number:  23246S

 Related applications:

Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Reagent is a quick and ready-to-use modification of the well-known Bradford coomassie dye-binding, colorimetric method for total protein quantitation. Proprietary additives to the Bradford Reagent make it compatible with up to 1% or higher of detergents and lysis reagents that are commonly used in life science research, including Triton® X-100 and NP-40.

• Convenient—detergent-free standard curve
• Flexible—compatible with samples both with and without detergent
• Minimal sample—requires only 10 µL for microplate procedure
• Colorimetric—measure with a standard spectrophotometer or plate reader (595 nm)
• Easy to use—single reagent, no working reagent preparation required
• Fast—10 minute incubation at room temperature
• Broad range—detects protein concentration in the ranges of 2 to 1500 µg/mL

Similar to the Bradford method, coomassie dye binds protein in an acidic medium causing an immediate shift in absorption maximum from 465 nm to 595 nm with a concomitant color change from green to blue. In addition, the assay is complete in just 10 minutes.

The protein assay can be performed in either test tube or microplate format. The standard working range is 100-1500 µg/mL with up to 1% detergent (or higher in some cases). Protein concentrations are estimated by reference to absorbances obtained for a series of standard protein dilutions, typically bovine serum albumin (BSA), which are assayed alongside the unknown samples. Because the color response with coomassie dye is non-linear with increasing protein concentration, a standard curve must be completed with each assay. Standards can be used directly without preparing them in the same detergent found in the test samples.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Range: 100-1,500µg/mL or 2-25µg/mL
Assay Type: Protein Quantitation Assay
Detection Method: Colorimetric (595 nm)
For Use With (Equipment): Microplate Reader, Spectrophotometer
Product Line: Pierce™
Product Size: 15 test tube assays
Technique: Absorbance, Solution-based Detection

Contents & storage

Detergent Compatible Bradford Assay Reagent, 25 mL, store at 4°C

Documents

Manuals & protocols