2,6-Dichloropyrazine, 99+%
2,6-Dichloropyrazine, 99+%
2,6-Dichloropyrazine, 99+%
Acros Organics

2,6-Dichloropyrazine, 99+%

CAS: 4774-14-5 | C4H2Cl2N2 | 148.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 232540050
also known as 23254
HSN 29339900
Price (INR)
13,747.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
13,747.00
Each
Chemical Identifiers
CAS4774-14-5
IUPAC Name2,6-dichloropyrazine
Molecular FormulaC4H2Cl2N2
InChI KeyLSEAAPGIZCDEEH-UHFFFAOYSA-N
SMILESClC1=CN=CC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point53°C to 56°C
GC>=99.0 %
Appearance (Color)White to beige
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only