Potassium hydroxide, ca. 85%, extra pure, flakes
Potassium hydroxide, ca. 85%, extra pure, flakes
Potassium hydroxide, ca. 85%, extra pure, flakes
Thermo Scientific Chemicals

Potassium hydroxide, ca. 85%, extra pure, flakes

CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 232550010
Price (USD)
58.65
Online Exclusive
64.90
Save 6.25 (10%)
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
58.65
Online Exclusive
64.90
Save 6.25 (10%)
Each
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Flakes
Titration with HCl>=85 %
Chloride (Cl)=<80 ppm
Iron (Fe)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only