Nunc™ Seals, Tapes, and Foils
Nunc™ Seals, Tapes, and Foils
Nunc™ Seals, Tapes, and Foils
Nunc™ Seals, Tapes, and Foils
Thermo Scientific™

Nunc™ Seals, Tapes, and Foils

Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialFor Use With (Application)Certified RNase/DNase-free
232698AluminumPCR, storage in 96-well platesYes
232699AluminumPCR, storage in 384 well platesYes
232701PolypropylenePCRYes
232702PolypropylenePCR, fluorescence assays, microscopyYes
235306PolyethyleneLow autofluorescence, ELISANo
235307PolyolefinPCR, qPCR, ELISA, Protein crystallographyYes
236272VinylLuminescence, photosensitive samplesNo
236366PolyesterCell culture, sterile applicationsNo
236366PKPolyesterCell culture, sterile applicationsNo
236370PolyesterGeneral purpose, sample detectionNo
236703VinylFluorescence, photosensitive samplesNo
236707PolyesterGeneral purpose, ELISAYes
236710Polyolefin AcrylateLow autofluorescenceYes
241205Non-woven RayonBreathable, Cell cultureNo
249720Non-woven RayonCell growth, Bacterial growthYes
250050PlasticConsistent sealNo
276014AluminumPCR, long-term storageYes
3501PolyesterShort-term storage, ELISANo
5701TS1PolyesterShort-term storage, ELISA, minimizes evaporation and cross contaminationNo
AB0718PolypropyleneVentilating and storing cell culturesNo
AB0580PVC-coated PolypropyleneCrystallography and genotypingNo
Catalog number 232698
Price (USD)
111.40
Online exclusive
124.03
Save 12.63
Case of 100
Material:
Aluminum
For Use With (Application):
PCR, storage in 96-well plates
Certified RNase/DNase-free:
Yes
Price (USD)
111.40
Online exclusive
124.03
Save 12.63
Case of 100

Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.

  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
AutoclavableNot Autoclavable
DescriptionAL SEAL TAPE FOR 96 WELL PLT
Evaporation BarrierMalleable allows for visual verification of seal security, minimizes evaporation and cross contamination
For Use With (Application)PCR, storage in 96-well plates
Incubation ConditionsGeneral
No. per Case100
No. per Pack100
PeelableNo
Product TypeAdhesive Seal
Solvent Or Alcohol ResistantLife Science Reagent Resistant
Temperature Range (Metric)-80°C to +120°C
Well ShapeAll well shapes
AdhesiveAcrylic
Certified RNase/DNase-freeYes
ColorFoil
MaterialAluminum
No. of Wells96
PierceableYes
Plate CompatibilityAll Microplates
Product LineNunc
SterilityNon-sterile
Unit SizeCase of 100
Contents & Storage
General; Long term

Frequently asked questions (FAQs)

Can I boil Nunc Sealing Tape?

Our Nunc Stealing Tape is thermostable from -20 degrees C to 100 degrees C.