Etilcelulosa, etoxil, contenido 48 %, 300 mPas
Etilcelulosa, etoxil, contenido 48 %, 300 mPas
Etilcelulosa, etoxil, contenido 48 %, 300 mPas
Thermo Scientific Chemicals

Etilcelulosa, etoxil, contenido 48 %, 300 mPas

CAS: 9004-57-3 | C23H24N6O4 | 448.483 g/mol
Have Questions?
Número de catálogoCantidadEnvase
2327000515 kgTambor de plástico
Número de catálogo 232700051
Precio (USD)
-
Cantidad:
5 kg
Envase:
Tambor de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS9004-57-3
IUPAC Name2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
Molecular FormulaC23H24N6O4
InChI KeyARSKJXYLLONUAJ-UHFFFAOYSA-N
SMILESCCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Viscosity250 to 350 cP (5 % in 80/20 toluene/ethanol at 25°C)
Appearance (Color)White
Appearance (Form)Granular powder
Assay48 to 49.5 % (ethoxyl)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only