D-Panthenol, 98+%
D-Panthenol, 98+%
D-Panthenol, 98+%
Thermo Scientific Chemicals

D-Panthenol, 98+%

CAS: 81-13-0 | C9H19NO4 | 205.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 232805000
Price (USD)
195.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
195.00
Each
Chemical Identifiers
CAS81-13-0
IUPAC Name(2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
Molecular FormulaC9H19NO4
InChI KeySNPLKNRPJHDVJA-ZETCQYMHSA-N
SMILESCC(C)(CO)[C@@H](O)C(=O)NCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+29.0° to +32.0° (20°C, 589 nm) (c=5, H2O)
Appearance (Form)Viscous liquid or solid
Refractive index1.4950 to 1.5020 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only