2-Pyrrolidone-5-carboxylic acid, sodium salt, 50 wt% solution in water
2-Pyrrolidone-5-carboxylic acid, sodium salt, 50 wt% solution in water
2-Pyrrolidone-5-carboxylic acid, sodium salt, 50 wt% solution in water
Thermo Scientific Chemicals

2-Pyrrolidone-5-carboxylic acid, sodium salt, 50 wt% solution in water

CAS: 54571-67-4 | C5H6NNaO3 | 151.10 g/mol
Quantity:
25 mL
100 mL
Catalog number 232920250
Price (JPY)
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS54571-67-4
SpecificationsSpecification SheetSpecification Sheet
Water47.5 to 52.5 % (K.F.)
pH6.8 to 7.4
Titration with HClO447.5 to 52.5 %
Appearance (Form)Solution
Appearance (Color)Colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only