Petroleum ether, extra pure, boiling range 100-140°C
Petroleum ether, extra pure, boiling range 100-140°C
Petroleum ether, extra pure, boiling range 100-140°C
Thermo Scientific Chemicals

Petroleum ether, extra pure, boiling range 100-140°C

CAS: 64742-49-0
Quantity:
10 L
25 L
Packaging:
View all
Plastic Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 233020100
Price (JPY)
-
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS64742-49-0
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
SpecificationsSpecification SheetSpecification Sheet
Benzene=<50 ppm
Boiling point98°C to 140°C
Color scale=<10 APHA
Refractive index1.4070 to 1.4150 (20°C, 589 nm)
Aromatics=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only