O-Methylisourea hemisulfate, 94%
O-Methylisourea hemisulfate, 94%
O-Methylisourea hemisulfate, 94%
Thermo Scientific Chemicals

O-Methylisourea hemisulfate, 94%

CAS: 52328-05-9 | 0·5 H2SO4 | 123.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
Catalog number 233032500
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS52328-05-9
IUPAC Namemethoxymethanimidamide
Molecular FormulaC2H6N2O
InChI KeyRMAHPRNLQIRHIJ-UHFFFAOYSA-N
SMILESCOC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Titration with NaOH>=92 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only