Chemical Identifiers
CAS4465-65-0
IUPAC Name4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-3,4,4a,5,12,12a-hexahydrotetracene-2-carboxamide hydrochloride
Molecular FormulaC22H23ClN2O7
InChI KeySPFAOPCHYIJPHJ-UHFFFAOYNA-N
SMILESCl.CN(C)C1C2CC3=C(C(O)=C4C(O)=CC=CC4=C3C)C(=O)C2(O)C(O)=C(C(N)=O)C1=O
Ver mais