Anhydrotetracycline hydrochloride, 'can be used as a secondary standard'
Anhydrotetracycline hydrochloride, 'can be used as a secondary standard'
Anhydrotetracycline hydrochloride, 'can be used as a secondary standard'
Thermo Scientific Chemicals

Anhydrotetracycline hydrochloride, 'can be used as a secondary standard'

Anhydrotetracycline hydrochloride, CAS # 13803-65-1, is a tetracycline derivative showing antibacterial activity commonly used to control tetracycline-dependent gene expression systems in bacteria.
Have Questions?
Catalog NumberQuantityPackaging
233131000100mgGlass bottle
Catalog number 233131000
Price (USD)
236.00
Each
Quantity:
100mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
236.00
Each
Chemical Identifiers
CAS13803-65-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Infrared spectrumConforms
Appearance (Form)Powder
Purity>=80 % (as is, calculated)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Anhydrotetracycline hydrochloride is a degradation product derived from tetracycline hydrochloride. It acts as an effector of tetracycline-dependent gene expression in bacterial expression systems. In this system the transcription of genes is reversibly turned on/off in the presence of several compounds including, anhydrotetracycline hydrochloride.
  • This compound poorly binds to the 30S ribosomal subunit compared with its precursor, tetracycline.

Applications

  • Anhydrotetracycline hydrochloride is commonly used in all fields of biology for control of transcription or translation processes
  • This compound has been used in laboratory experiments and shows lower antibacterial activity against Escherichia coli compared with tetracycline.
RUO – Research Use Only