Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
Thermo Scientific Chemicals

Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 233281000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-30-8
IUPAC Namepentanedial
Molecular FormulaC5H8O2
InChI KeySXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILESO=CCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific gravity(20/20°C) 1.000 to 1.133
Appearance (Form)Clear to slightly hazy solution
Assay25 to 27 % w/w
pH3.1 to 4.5 (at 25°C)
Color scale=<100 APHA
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only