RPMI 1640 Medium, powder, HEPES
Gibco™

RPMI 1640 Medium, powder, HEPES

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
2340002110 x 1 L
2340006210 L
Número do catálogo 23400021
Preço (BRL)
802,35
Each
Quantity:
10 x 1 L
Customize this product
Preço (BRL)
802,35
Each

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes and carcinomas. Life Technologies offers a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications.

This RPMI is modified as follows:

With: L-glutamine, Phenol Red, HEPES

Without: Sodium Bicarbonate

The complete formulation is available.

Gibco™ RPMI 1640 is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12 and PABA which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations.

Product Intended Use

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

RPMI 1640 contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Please note that the shelf life on delivered products varies. This product has a minimum shelf life of 6 months at time of shipment. For more information regarding product shelf life, including details on specific product lots, please contact Customer Service.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificações
Cell LineHeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell TypeLeukemic Cells
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeRPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Quantity10 x 1 L
Shelf Life36 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormPowder
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesGlutamine, HEPES, Phenol Red
Without AdditivesNo Sodium Pyruvate, No Sodium Bicarbonate
Unit SizeEach
Conteúdo e armazenamento
Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture

Frequently asked questions (FAQs)

How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?

While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the density (g/L) for RPMI 1640 Medium?

We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the shelf life of my powdered media once I reconstitute it?

Expiration date of most Gibco reconstituted dry format media (AGT or DPM) has not been established; end users should assess performance and stability of this reconstituted media in a system that is relevant to their process. A dry format product (either DPM or AGT) will age upon storage and while we may not be able to detect which component(s) degrade since we can't measure every component, that doesn't mean that the reconstituted liquid will have the same stability as a fresh liquid made by direct weigh methods.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.