Pierce™ Far-Western Blot Kit for Biotinylated Proteins
Thermo Scientific™

Pierce™ Far-Western Blot Kit for Biotinylated Proteins

The Thermo Scientific Pierce Far-Western Blot Kit for Biotinylated Proteins enables in-gel or on-membrane Western blot detection of target proteinsRead more
Have Questions?
Catalog NumberQuantity
23500220 mL
Catalog number 23500
Price (USD)
760.00
Each
Quantity:
220 mL
Request bulk or custom format
Price (USD)
760.00
Each
The Thermo Scientific Pierce Far-Western Blot Kit for Biotinylated Proteins enables in-gel or on-membrane Western blot detection of target proteins via interaction with any biotin-labeled test protein used as the primary probe.

Features of the Far-Western Blot Kit for Biotinylated Proteins:

• On-membrane or in-gel detection options—on-membrane detection offers greater sensitivity; in-gel detection is faster and prevents problems associated with incomplete or inefficient transfer
• Nonradioactive alternative for far-Western analysis—reliable and sensitive biotin/streptavidin-HRP chemistry combined with chemiluminescent detection offers a practical and safe alternative to radiolabeling the bait protein
• Useful interaction range—effective for interaction pairs that have moderate to strong binding associations between the prey and the biotinylated bait protein
• Primary antibody-free detection—uses a biotinylated protein as the probe, eliminating the need for primary antibodies or even to have prior knowledge of target-protein identities
• Compatible with both SDS-PAGE and native gels—provides option to probe for prey proteins in a more native environment because reduced or denaturing systems may not always allow for the intended interaction
• Reduced nonspecific binding—biotin/streptavidin-HRP systems demonstrate less nonspecific binding compared to antibodies directed against the bait protein
• Compatible with protein staining—gels can be used for total protein staining after the in-gel chemiluminescent detection step, eliminating the need to run two gels

This far-Western blotting kit provides the reagents and protocol necessary to probe for target proteins by virtue of their interaction with a specific binding partner, which the researcher supplies in purified, biotinylated form. The kit is successful for analysis of protein-protein interactions in which the target (prey) proteins can be bound after they have been separated by gel electrophoresis. The kit includes reagents and procedures for probing and chemiluminescent detection with blots or directly from mini gels.

Includes:
• Streptavidin-HRP conjugate, dilution and wash buffers, chemiluminescent substrate solutions, and cellophane sheets for mini gels

Applications:
• Protein interaction discovery—search and screen for proteins in cell lysates that have binding affinity to the purified biotinylated protein
• Protein interaction confirmation—characterize and identify the presence of native or off-target binding interactions involving a given biotinylated protein and particular samples

Far-Western blotting involves probing for targets first with a putative interaction-protein rather than a primary antibody. The method is called far-Western blotting because primary antibodies are used secondarily to detect the primary interaction probe rather than the target protein. The Pierce Far-Western Blot Kit for Biotinylated Proteins eliminates the use of antibodies altogether. The primary probe (i.e., the biotinylated bait protein supplied by the researcher) is secondarily detected with streptavidin-HRP conjugate, whose activity is visualized by reaction with a very sensitive chemiluminescent substrate.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ReactivityAll species
Sufficient For10 Mini Gels
Detection MethodChemiluminescence
Product LinePierce
Product TypeWestern Blot Kit
Quantity220 mL
SubstrateStreptavidin-HRP
Unit SizeEach
Contents & Storage
Sufficient For: 10 mini gel or blot experiments with a biotinylated-protein probe
• Streptavidin-HRP Conjugate, 0.1 mg
• Dilution Buffer (10X), 50 mL
• PBS Packs (each makes 500 mL), 17 packs
• Luminol Enhancer, 55 mL
• Stable Peroxide Buffer, 55 mL
• Surfact-Amps 20 Detergent Solution, 6 x 10 mL
• Pre-Cut Cellophane, 10 sheets

Store kit at 4°C.

Frequently asked questions (FAQs)

If my protein is endogenously expressed, which Thermo Scientific Protein:Protein Interaction (PPI) kit should I choose?

We recommend the biotin kit for endogenously expressed proteins. The reason for this is the wide variety of biotinylation reagents that can be used to label the detection protein of the binding pair. For help choosing a biotinylation reagent, refer to the Biotinylation Selection Guide available at our website.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

How is the tagged protein detected on the membrane using the Thermo Scientific Protein:Protein Interaction (PPI) kits?

The biotin kit utilizes streptavidin-HRP to detect the biotinylated protein bound to the prey protein on the membrane. The GST kit utilizes a polyclonal goat anti-GST antibody for detection of the glutathione-S-transferase tagged fusion protein bound to another protein on a membrane.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

How many reactions can be performed using one Thermo Scientific Protein:Protein Interaction (PPI) kit?

The Thermo Scientific Pull-Down Protein:Protein Interaction Kits contain a complete, validated set of reagents specifically developed for performing pull-down assays.The kits contain sufficient materials for conducting 25 pull-down assays.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Are all of the conditions for binding the prey protein to the bait protein described in the Thermo Scientific Protein:Protein Interaction (PPI) kits?

No. Every protein pair will require unique conditions in order to bind with one another. Factors to consider are pH, ionic strength of the buffer, need for cofactors, whether the interaction is long term or transient, etc. The researcher is recommended to perform a literature search or try several different conditions in order to determine what conditions are best for the binding of their proteins of interest. The kit includes a “universal“ wash buffer formed by diluting the lysis buffer 1:1 in TBS.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

With the Thermo Scientific Protein:Protein Interaction (PPI) kits, what methods can be used to detect the prey protein?

The prey protein can be detected following transfer on a membrane. The membrane procedures uses 1:100,000 - 1:500,000 dilution of the anti-GST antibody.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.