1,4-Dioxaspiro[4.5]decan-8-one, 98%
1,4-Dioxaspiro[4.5]decan-8-one, 98%
1,4-Dioxaspiro[4.5]decan-8-one, 98%
Acros Organics

1,4-Dioxaspiro[4.5]decan-8-one, 98%

CAS: 4746-97-8 | C8H12O3 | 156.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
235640250
also known as 23564
25 gGlass bottle
Catalog number 235640250
also known as 23564
HSN 29329900
Price (INR)
14,953.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
14,953.00
Each
Chemical Identifiers
CAS4746-97-8
IUPAC Name1,4-dioxaspiro[4.5]decan-8-one
Molecular FormulaC8H12O3
InChI KeyVKRKCBWIVLSRBJ-UHFFFAOYSA-N
SMILESO=C1CCC2(CC1)OCCO2
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point70°C to 74°C
GC>=97.5 %
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only