Search
| Número do catálogo | Product Type | Holds |
|---|---|---|
| 236011 | Flask Clamp | 50 mL Erlenmeyer Flask |
| 236012 | Flask Clamp | 125 mL Erlenmeyer Flask |
| 236013 | Flask Clamp | 250/300 mL Erlenmeyer Flask |
| 236014 | Flask Clamp | 500 mL Erlenmeyer Flask |
| 236015 | Flask Clamp | 1 L Erlenmeyer Flask |
| 236016 | Flask Clamp | 2 L Erlenmeyer Flask |
| 236019 | Flask Clamp | 2800 mL Fernbach Flask |
| 236025 | Microplate Clamp | Microplates |
| 236099 | Flask Clamp Kit | Flasks |
Secure vessels using Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers. With exceptional innovation, reliability and accuracy, these orbital shakers, platforms and accessories are the optimal choice for a busy lab.
Note:
Replacement screws also available separately.