Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Thermo Scientific™
Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Secure vessels using these universal platform clamps for orbital shakers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeHolds
236025Microplate ClampMicroplates
236011Flask Clamp50 mL Erlenmeyer Flask
236012Flask Clamp125 mL Erlenmeyer Flask
236013Flask Clamp250/300 mL Erlenmeyer Flask
236014Flask Clamp500 mL Erlenmeyer Flask
236015Flask Clamp1 L Erlenmeyer Flask
236016Flask Clamp2 L Erlenmeyer Flask
236019Flask Clamp2800 mL Fernbach Flask
236099Flask Clamp KitFlasks
Catalog number 236025
Price (BRL)
-
Product Type:
Microplate Clamp
Holds:
Microplates
Secure vessels using Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers. With exceptional innovation, reliability and accuracy, these orbital shakers, platforms and accessories are the optimal choice for a busy lab.
Flask Clamp Starter Kit includes:
  • (1) microplate clamp
  • (2) 25mL flask clamps
  • (2) 50mL flask clamps
  • (2) 125mL flask clamps
  • (4) 250/300mL flask clamps
  • (4) 500mL flask clamps
  • (2) 1L flask clamps
  • (2) 2L flask clamps
  • Replacement screws
Note:
Replacement screws also available separately.
Specifications
DescriptionMicroplate Clamp
For Use With (Equipment)MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
HoldsMicroplates
Product TypeMicroplate Clamp
Unit SizeEach