MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps
MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps
Actual product may vary
MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps
MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps
Thermo Scientific™
MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps
Maximize capacity when working with a single-size vessel by using HP Orbital Shakers Dedicated Platforms with pre-installed clamps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsClampsFor Use With (Equipment)
238050250/300 mL Erlenmeyer Flask25MaxQ HP 416/420 Shaker
238019125 mL Erlenmeyer Flask61MaxQ HP 430/435/436/480/481 Shaker
Catalog number 238050
Price (MXN)
-
Request A Quote
Holds:
250/300 mL Erlenmeyer Flask
Clamps:
25
For Use With (Equipment):
MaxQ HP 416/420 Shaker
Maximize capacity when working with a single-size vessel by using Thermo Scientific™ HP Orbital Shakers Dedicated Platforms with pre-installed clamps.
Specifications
Clamps25
Dimensions (L x W)18 x 18 in.
For Use With (Equipment)MaxQ HP 416/420 Shaker
Holds250/300 mL Erlenmeyer Flask
Length (English) Platform18 in.
Length (Metric) Platform45.7 cm
Product TypeShaker Platform with Clamps
Width (English) Platform18 in.
Width (Metric) Platform45.7 cm
Unit Size-