Agarosa, para bioquímica, polvo puro (MR = -0.17)
Agarosa, para bioquímica, polvo puro (MR = -0.17)
Agarosa, para bioquímica, polvo puro (MR = -0.17)
Thermo Scientific Chemicals

Agarosa, para bioquímica, polvo puro (MR = -0.17)

CAS: 9012-36-6
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
500 g
Envase:
Frasco de vidrio
Número de catálogo 239025000
Precio (EUR)
1.037,00
Each
Cantidad:
500 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
1.037,00
Each
Identificadores químicos
CAS9012-36-6
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC24H38O19
InChI KeyMJQHZNBUODTQTK-WKGBVCLCSA-N
SMILESOC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Loss on drying=<7 % (5 g, 110°C, 24 h)
Gel point34.5°C to 37.5°C (1.5 % gel)
Appearance (Form)Powder
Ash=<0.5 %
Gel strength>=2000 g/cm² (1.5 % gel)
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only