Nalgene™ LDPE Economy Wash Bottles
Nalgene™ LDPE Economy Wash Bottles
Nalgene™ LDPE Economy Wash Bottles
Nalgene™ LDPE Economy Wash Bottles
Thermo Scientific™

Nalgene™ LDPE Economy Wash Bottles

Prevent separation when squeezing these LDPE economy wash bottles, which feature a tight-fitting draw tube and closure.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoCapacity (Metric)Closure Size
2401-0500500 mL28 mm
2401-0125125 mL24 mm
2401-0250250 mL24 mm
2401-10001000 mL38-430 mm
Número do catálogo 2401-0500
Preço (BRL)
-
Capacity (Metric):
500 mL
Closure Size:
28 mm

High quality squeeze bottles at a value price. Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.

Features:

  • Polypropylene screw closure/stem with PPCO draw tube
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Sputter-free, precision stream
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.

Especificações
Capacity16 oz., 500 mL
Capacity (English)16 oz.
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Diameter (Metric)91.4 mm
Height (English)8.4 in.
Height (Metric)213 mm
IncludesStem and draw tube
No. per Case24
No. per Pack6
Product TypeEconomy Wash Bottle
ShapeCylindrical
Through the Closure DispensingYes
Capacity (Metric)500 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialLDPE
MouthNarrow
Unit SizeCase of 24