Search
View additional product information for EBSS, calcium, magnesium, phenol red - FAQs (24010043)
2 product FAQs found
Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Phenol red in tissue culture is a weak estrogen. Implications concerning study of estrogen-responsive cells in culture are discussed in Proc Natl Acad Sci U S A 83:2496 (1986).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.