Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
Thermo Scientific™

Nalgene™ Wash Bottles made with Teflon™ fluoropolymer

Ensure contamination-free dispensing with these chemical-resistant wash bottles made with Teflon™ fluoropolymer.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoCapacity (Metric)Closure Size
2403-0250250 mL24 mm
2403-0125125 mL24 mm
2403-0500500 mL38-430 mm
2403-10001000 mL38-430 mm
Número do catálogo 2403-0250
Preço (BRL)
-
Capacity (Metric):
250 mL
Closure Size:
24 mm
Ensure contamination-free dispensing with Thermo Scientific™ Nalgene™ Wash Bottles made with Teflon™ fluoropolymer, useful for trace element work and other high-purity applications. These phthalate-free bottles offer excellent chemical resistance and are molded entirely of fluoropolymer. Designed to dispense through the top with a closure and stem molded in one piece for reliable service. The precision-molded tip provides a straight, steady stream and ultimate control without sputtering.
  • With ETFE screw closure/stem and draw tube
  • Demonstrates excellent chemical resistance to nearly any chemical in your lab including solvents—won't become brittle or crack
  • Inert fluoropolymer construction is low in extractables and low binding—useful for trace element work
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Autoclavable/Transparent
  • Sputter-free, precision stream
.

Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.

Especificações
AutoclavableAutoclavable
Capacity8 oz., 250 mL
Capacity (English)8 oz.
Closure MaterialETFE
Closure Size24 mm
Closure TypeScrew Cap
Diameter (Metric)60.2 mm
Height (English)7 in.
Height (Metric)178 mm
Product TypeWash Bottle
ShapeCylindrical
Through the Closure DispensingYes
Capacity (Metric)250 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialFEP
MouthNarrow
Unit SizeCase of 4

Frequently asked questions (FAQs)

Why do you recommend Teflon-coated bottles to store high-value contents such as, active pharmaceutical ingredients (API's), bulk intermediates, and critical reagents?

Teflon is ideal for storing the highest-value contents because it is resistant to most chemicals, temperatures, and processes, allowing for rigorous cleaning procedures such as those that require boiling nitric acid. Teflon also allows you to store contents down to cryogenic temperatures for products requiring ultra-low temperature (below -100 degrees C) storage.
We offer Nalgene Teflon bottles that provide excellent chemical and temperature resistance in a leak-proof design. They are manufactured using quality resins, with the added value of high-level support, making Nalgene Teflon bottles an ideal choice for your high-value contents.