Nalgene™ PPCO Wash Bottles, 500mL - FAQs
View additional product information for Nalgene™ Autoclavable PPCO Wash Bottles - FAQs (2405-1000, 2405-1000PK, 2405-0500, 2405-0500PK)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.