Nalgene™ Autoclavable PPCO Wash Bottles
Nalgene™ Autoclavable PPCO Wash Bottles
Nalgene™ Autoclavable PPCO Wash Bottles
Nalgene™ Autoclavable PPCO Wash Bottles
Nalgene™ Autoclavable PPCO Wash Bottles
Thermo Scientific™

Nalgene™ Autoclavable PPCO Wash Bottles

Reduce the risk of contamination when dispensing sterile water or reagents by using these autoclavable PPCO wash bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2405-0500PK500 mL28 mmPack of 6
2405-0500500 mL28 mmCase of 24
2405-10001000 mL38-430 mmCase of 12
2405-1000PK
also known as 2405-1000
1000 mL38-430 mmPack of 4
Catalog number 2405-0500PK
Price (USD)
91.57
Online exclusive
101.70
Save 10.13
Pack of 6
Capacity (Metric):
500 mL
Closure Size:
28 mm
Unit Size:
Pack of 6
Price (USD)
91.57
Online exclusive
101.70
Save 10.13
Pack of 6

Reduce the risk of contamination when dispensing sterile water or reagents by using autoclavable Thermo Scientific™ Nalgene™ PPCO Wash Bottles. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. These squeeze bottles are flexible, contact-clear, and designed to dispense through the top with a closure and stem molded in one piece for reliable service. Bent stem dispensing tube retains its bend after autoclaving but bend angle can be modified by hand while warm if desired. The tight fit of the draw tube and closure prevents separation when squeezing bottle.

  • Autoclavable
  • Polypropylene screw/stem and draw tube
  • Tip can be cut back to provide increased flow
  • Sputter-free, precision stream
  • Bottle and Carton are recyclable in many communities
  • Manufactured from non-toxic materials

Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.

Specifications
AutoclavableAutoclavable
Capacity16 oz., 500 mL
Capacity (English)16 oz.
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeCap
Diameter (Metric)72.6 mm
Height (English)8.4 in.
Height (Metric)213 mm
IncludesScrew/stem and draw tube
No. per Case24
No. per Pack6
Product TypeWash Bottle
ShapeCylindrical
Through the Closure DispensingYes
Capacity (Metric)500 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialPPCO
MouthNarrow
SterilityNon-sterile
Unit SizePack of 6

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.