KingFisher Apex Accessories
Imagem ilustrativa. O produto pode variar
KingFisher Apex Accessories
KingFisher Apex Accessories
KingFisher Apex Accessories
KingFisher Apex Accessories
KingFisher Apex Accessories
KingFisher Apex Accessories
Thermo Scientific™

KingFisher Apex Accessories

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoProduct Type
24075960Heating Block 24
24075970Matrix Heating Block
2407991096 PCR Head
24079920Combi Head
2407993096 DW Head
2407994024 Combi Head
24075910PCR Plate Heating Block
2407592096 KF Heating Block
2407593096 DW Heating Block
2407594024 DW Heating Block
24075950Heating Block 96
N21445Storage Tube Adapter
N21446Plate Adapter
N21447UV Lamp (Replacement)
Número do catálogo 24075960
Preço (BRL)
-
Product Type:
Heating Block 24
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality
  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
Especificações
DisposableReusable
For Use With (Equipment)KingFisher Apex
Quantity1 Pc.
Capacity24 Samples
Format24
Product TypeHeating Block 24
Unit SizeEach