KF Apex 96 DW head - FAQs
View additional product information for KingFisher Apex Accessories - FAQs (24075960, 24075970, 24079910, 24075910, 24079940, 24075920, 24075930, 24079920, 24075940, 24079930, 24075950, N21446, N21445, N21447)
1 product FAQs found
How many kinds (type or size) of magnets are there for the KingFisher Apex 96 DW Head (Cat. No. 24079930)?
Please see the Thermo Scientific KingFisher Flex User Manual, page 32. As you will see, there are four different heads. In terms of magnet force, it's a permanent magnet so the expected life is >100 years.