Sesame oil
Acros Organics

Sesame oil

CAS: 8008-74-0
Have Questions?
Catalog NumberQuantityPackaging
241002500250 mLGlass bottle
Catalog number 241002500
HSN 29420090
Price (INR)
8,562.00
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,562.00
Each
Chemical Identifiers
CAS8008-74-0
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Acid value=<0.6 mg KOH/g
Appearance (Form)Oily liquid
Refractive index1.4700 to 1.4760 (20°C, 589 nm)
Peroxide value=<10 meq/kg
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only