L(-)-Carnitine, 99+%
L(-)-Carnitine, 99+%
L(-)-Carnitine, 99+%
Thermo Scientific Chemicals

L(-)-Carnitine, 99+%

CAS: 541-15-1 | C7H15NO3 | 161.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 241040010
Price (TWD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS541-15-1
IUPAC Name3-hydroxy-4-(trimethylazaniumyl)butanoate
Molecular FormulaC7H15NO3
InChI KeyPHIQHXFUZVPYII-UHFFFAOYNA-N
SMILESC[N+](C)(C)CC(O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration with HClO4>=99.0 %
Specific optical rotation-29° to -32° (20°C, 589 nm) (c=1, H2O)
Solubility(5 % in water) Clear colorless
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only