8M Guanidine-HCl Solution

Catalog number: 24115B

Thermo Scientific™  Related applications: Protein Labeling & Crosslinking | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce 8M Guanidine-HCl Solution is an accurately formulated, ready-to-use solution of guanidine hydrochloride, that can be easily diluted and pH-adjusted to any concentration below 8M.

Features of 8M Guanidine-HCl Solution:

• Any concentration below 8M guanidine can be quickly and easily prepared.
• No wasting valuable research time weighing and dissolving guanidine crystals
• Free from UV absorbing materials in the range of 225-300nm
• Free from heavy metal contaminants
• Free from degradation products such as ammonia
• Excellent stability

Applications of Guanidine Hydrochloride:
• Solubilizing proteins from inclusion bodies
• Increasing solubility of hydrophobic peptides and proteins
• Denaturing proteins

Chaotropes, substances that disrupt the structure of water interactions, help to solubilize hydrophobic proteins and peptides for a variety of biological purposes. For example, hydrophobic peptides and proteins may be dissolved in coupling buffer containing either 6M guanidine-HCl or 4M urea prior to immobilization on AminoLink Plus Support to create an affinity column. However, because the degradation products of guanidine and urea include ammonia, which contains a primary amine that will compete for reaction to the support, urea and guanidine solutions must be prepared with high quality reagents immediately before use.

Guanidine is a general protein denaturant, unfolding proteins and altering their three-dimensional structure. Consequently, some proteins will be irreversibly altered upon interaction with guanidine solutions and may lose their binding function. Before any large-scale use of guanidine, it is best to test a small sample and determine whether the denaturing effects will adversely affect the intended use of the protein.

Related Products
Guanidine-HCl
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Size: Custom
Reagent Type: Chaotrope, Denaturant

Contents & storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.

Documents

Manuals & protocols