Phloroglucinol, 99+%, anhydrous
Phloroglucinol, 99+%, anhydrous
Phloroglucinol, 99+%, anhydrous
Thermo Scientific Chemicals

Phloroglucinol, 99+%, anhydrous

CAS: 108-73-6 | C6H6O3 | 126.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 241765000
Price (USD)
348.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
348.00
Each
Chemical Identifiers
CAS108-73-6
IUPAC Namebenzene-1,3,5-triol
Molecular FormulaC6H6O3
InChI KeyQCDYQQDYXPDABM-UHFFFAOYSA-N
SMILESOC1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<2 % (1 g, 120°C)
Appearance (Color)White to light yellow to light beige
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point213°C to 221°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only