2-chloro-5-(trifluoromethyl)pyridine, 98+%
2-chloro-5-(trifluoromethyl)pyridine, 98+%
2-chloro-5-(trifluoromethyl)pyridine, 98+%
2-chloro-5-(trifluoromethyl)pyridine, 98+%
Thermo Scientific Chemicals

2-chloro-5-(trifluoromethyl)pyridine, 98+%

CAS: 52334-81-3 | C6H3ClF3N | 181.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 241781000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52334-81-3
IUPAC Name2-chloro-5-(trifluoromethyl)pyridine
Molecular FormulaC6H3ClF3N
InChI KeyJFZJMSDDOOAOIV-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystalline solid
Infrared spectrumConforms
Melting point29°C to 33°C
GC>=98.0 %
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only