L(-)-Glucose, 98%
L(-)-Glucose, 98%
L(-)-Glucose, 98%
Thermo Scientific Chemicals

L(-)-Glucose, 98%

CAS: 921-60-8 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 241922500
Price (USD)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS921-60-8
IUPAC Name6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyWQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILESOCC1OC(O)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-49° to -54° (20°C, 589 nm) (c=1, water, NH3)
Appearance (Color)White to almost white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only