Nalgene™ Color-Coded LDPE Wash Bottles
Nalgene™ Color-Coded LDPE Wash Bottles
Nalgene™ Color-Coded LDPE Wash Bottles
Nalgene™ Color-Coded LDPE Wash Bottles
Thermo Scientific™

Nalgene™ Color-Coded LDPE Wash Bottles

Simplify content identification with these narrow-mouth wash bottles that feature colored closures with an integrated stem.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure Color
2422-3500Blue
2422-2500Yellow
2422-5500Red
2422-4500Green
Catalog number 2422-3500
Price (CLP)
-
Closure Color:
Blue

High quality squeeze bottles with color-coded closures for easy identification. Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.

  • Color-coded top allows you to coordinate reagents with industry storage codes.
  • Tip can be cut back to provide increased flow.
  • Tight fit of draw tube and closure prevents separation when squeezing bottle.
  • Flexible LDPE bottle provides soft, ergonomic squeeze.
  • Sputter-free, precision stream.
  • Bottle and Carton are readily recyclable in most communities.
  • Manufactured from non-toxic materials.

Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.

Specifications
AutoclavableNot Autoclavable
Capacity (English)16 oz.
Closure ColorBlue
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
DescriptionWith stem and draw tube
Diameter (Metric)72.9 mm
Height (English)8.8 in.
Height (Metric)224 mm
No. per Case24
No. per Pack6
Product TypeColor-coded Wash Bottle
ShapeCylindrical
Through the Closure DispensingYes
Capacity (Metric)500 mL
Closure IncludedYes
ColorNatural
GraduatedNo
MaterialLDPE
MouthNarrow
Unit SizeCase of 24