Search
| Catalog Number | Closure Color | Chemical Label | Unit Size |
|---|---|---|---|
| 2428-0503PK also known as 2428-0503 | Green | Methanol | Pack of 6 |
| 2428-0507 | Natural | Deionized Water | Case of 24 |
| 2428-0507PK also known as 2428-0507 | Natural | Deionized Water | Pack of 6 |
| 2428-0504 | Yellow | Isopropanol | Case of 24 |
| 2428-0505 | Natural | Distilled Water | Case of 24 |
| 2428-0503 | Green | Methanol | Case of 24 |
| 2428-0502 | White | Ethanol | Case of 24 |
| 2428-0502PK also known as 2428-0502 | White | Ethanol | Pack of 6 |
| 2428-0506 | White | Sodium Hypochlorite | Case of 24 |
| 2428-0504PK also known as 2428-0504 | Yellow | Isopropanol | Pack of 6 |
| 2428-0506PK also known as 2428-0506 | White | Sodium Hypochlorite | Pack of 6 |
| 2428-0505PK also known as 2428-0505 | Natural | Distilled Water | Pack of 6 |
Prevent chemical accidents in the lab with Nalgene™ Right-to-Understand LDPE Wash Bottles. Bottles are pre-labeled with chemical identification and safe handling information following the Globally Harmonized System (GHS) specifications as applicable to secondary laboratory containers. Wash bottles are ready to use when you transfer common laboratory chemicals from manufacturers' containers. Self-venting wash bottles eliminate risk of bottles self-dispensing.