Nalgene™ Right-to-Understand Safety Wash Bottles featuring Globally Harmonized System (GHS) labeling

Catalog number: 2428-0506PK
Have Questions?

Nalgene™ Right-to-Understand Safety Wash Bottles featuring Globally Harmonized System (GHS) labeling

Catalog number: 2428-0506PK
Have Questions?
Prevent chemical accidents in the lab with Nalgene™ Right-to-Understand LDPE Wash Bottles. Bottles are pre-labeled with chemical identification and safe handling information following the Globally Harmonized System (GHS) specifications as applicable to secondary laboratory containers. Wash bottles are ready to use when you transfer common laboratory chemicals from manufacturers’ containers. Self-venting wash bottles eliminate risk of bottles self-dispensing.
 
Catalog Number
2428-0506PK
also known as 2428-0506
Unit Size
Pack of 6
Closure Color
White
For Use With (Application)
Sodium Hypochlorite
Price (USD)
Availability
-
Full specifications
Quantity
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
For Use With (Application)Sodium Hypochlorite
MaterialLDPE
PackagingPack of 6
Closure ColorWhite
Certifications/ComplianceGHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialHDPE
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePack of 6
Catalog Number
Unit Size
Case of 24
Closure Color
Natural
For Use With (Application)
Deionized Water
Price (USD)
Availability
-
Full specifications
Quantity
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Deionized Water
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorNatural
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
Catalog Number
2428-0507PK
also known as 2428-0507
Unit Size
Pack of 6
Closure Color
Natural
For Use With (Application)
Deionized Water
Price (USD)
Availability
-
Full specifications
Quantity
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Deionized Water
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorNatural
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePack of 6
Catalog Number
Unit Size
Case of 24
Closure Color
Yellow
For Use With (Application)
Isopropanol
Price (USD)
Availability
-
Full specifications
Quantity
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Isopropanol
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorYellow
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
Catalog Number
Unit Size
Case of 24
Closure Color
Natural
For Use With (Application)
Distilled Water
Price (USD)
Availability
-
Full specifications
Quantity
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Distilled Water
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorNatural
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
Catalog Number
Unit Size
Case of 24
Closure Color
Green
For Use With (Application)
Methanol
Price (USD)
Availability
-
Full specifications
Quantity
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Methanol
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorGreen
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
Catalog Number
2428-0503PK
also known as 2428-0503
Unit Size
Pack of 6
Closure Color
Green
For Use With (Application)
Methanol
Price (USD)
Availability
-
Full specifications
Quantity
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Methanol
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorGreen
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePack of 6
Catalog Number
Unit Size
Case of 24
Closure Color
White
For Use With (Application)
Ethanol
Price (USD)
Availability
-
Full specifications
Quantity
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Ethanol
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorWhite
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
Catalog Number
2428-0502PK
also known as 2428-0502
Unit Size
Pack of 6
Closure Color
White
For Use With (Application)
Ethanol
Price (USD)
Availability
-
Full specifications
Quantity
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Ethanol
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorWhite
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePack of 6
Catalog Number
Unit Size
Case of 24
Closure Color
White
For Use With (Application)
Sodium Hypochlorite
Price (USD)
Availability
-
Full specifications
Quantity
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorWhite
DescriptionWith stem and draw tube
For Use With (Application)Sodium Hypochlorite
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorWhite
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialHDPE
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
Showing 10 of 11Show more
Catalog NumberSpecificationsUnit SizeClosure ColorFor Use With (Application)Price (USD)AvailabilityQuantity
2428-0506PK
also known as 2428-0506
Full specifications
Pack of 6WhiteSodium Hypochlorite
-
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
For Use With (Application)Sodium Hypochlorite
MaterialLDPE
PackagingPack of 6
Closure ColorWhite
Certifications/ComplianceGHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialHDPE
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePack of 6
2428-0507Full specifications
Case of 24NaturalDeionized Water
-
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Deionized Water
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorNatural
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
2428-0507PK
also known as 2428-0507
Full specifications
Pack of 6NaturalDeionized Water
-
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Deionized Water
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorNatural
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePack of 6
2428-0504Full specifications
Case of 24YellowIsopropanol
-
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Isopropanol
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorYellow
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
2428-0505Full specifications
Case of 24NaturalDistilled Water
-
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Distilled Water
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorNatural
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
2428-0503Full specifications
Case of 24GreenMethanol
-
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Methanol
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorGreen
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
2428-0503PK
also known as 2428-0503
Full specifications
Pack of 6GreenMethanol
-
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Methanol
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorGreen
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePack of 6
2428-0502Full specifications
Case of 24WhiteEthanol
-
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Ethanol
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorWhite
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
2428-0502PK
also known as 2428-0502
Full specifications
Pack of 6WhiteEthanol
-
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Ethanol
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorWhite
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePack of 6
2428-0506Full specifications
Case of 24WhiteSodium Hypochlorite
-
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorWhite
DescriptionWith stem and draw tube
For Use With (Application)Sodium Hypochlorite
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorWhite
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialHDPE
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24
Showing 10 of 11Show more
  • Bottles are made from high quality Nalgene materials for low extractables and long life in the lab
  • Printed with 100% Globally Harmonized System (GHS) chemical safety information.  No other system markings are used on the bottles, which eliminates confusion
  • Color coded for easy chemical  identification at a glance
  • Dispensing nozzles are carefully molded to deliver a straight stream for precision dispensing and safety
  • Self-venting, so contents stay in bottle with no risk of leakage
  • Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof service
Right-to-Understand Safety Wash Bottles for acetone and other organic solvents are available in FEP (part number 2429-0501) for better chemical compatibility and product longevity.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text