Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Keep hazardous volatile liquids off the lab bench with Thermo Scientific™ Nalgene™ Vented Unitary Right-To-Know LDPE Wash Bottles, featuring a unique vent system. Unitary bottle prevents contamination when filling the bottle; no draw tube assembly to pick up contamination off the lab bench. A strong, steady, no sputter stream of liquid is dispensed when the wash bottle is squeezed. Marked with four color, NFPA diamonds, GHS compliant hazard symbols, and right-to-know chemical information. Choose from seven chemical markings.
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Through the Closure Dispensing | No |
Chemical Label | Distilled Water |
Product Type | Wash Bottle |
Capacity | 32 oz., 1000 mL |
Capacity (Metric) | 1000 mL |
Capacity (English) | 32 oz. |
Color | Translucent |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 12 |
No. per Pack | 2 |
Closure Color | Natural |
Unitary | Yes |
Certifications/Compliance | Meet OSHA requirements for workplace chemical container labeling. |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Unit Size | Pack of 2 |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Through the Closure Dispensing | No |
Chemical Label | Methanol |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Color | Green |
Unitary | Yes |
Certifications/Compliance | Meet OSHA requirements for workplace chemical container labeling. |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Unit Size | Case of 24 |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Through the Closure Dispensing | No |
Chemical Label | Methanol |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Color | Green |
Unitary | Yes |
Certifications/Compliance | Meet OSHA requirements for workplace chemical container labeling. |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Unit Size | Pack of 4 |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Through the Closure Dispensing | No |
Chemical Label | Isopropanol |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Color | Yellow |
Unitary | Yes |
Certifications/Compliance | Meet OSHA requirements for workplace chemical container labeling. |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Unit Size | Case of 24 |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Through the Closure Dispensing | No |
Chemical Label | Isopropanol |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Color | Yellow |
Unitary | Yes |
Certifications/Compliance | Meet OSHA requirements for workplace chemical container labeling. |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Unit Size | Pack of 4 |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Through the Closure Dispensing | No |
Chemical Label | Distilled Water |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Color | Natural |
Unitary | Yes |
Certifications/Compliance | Meet OSHA requirements for workplace chemical container labeling. |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Unit Size | Case of 24 |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Through the Closure Dispensing | No |
Chemical Label | Distilled Water |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Color | Natural |
Unitary | Yes |
Certifications/Compliance | Meet OSHA requirements for workplace chemical container labeling. |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Unit Size | Pack of 4 |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Through the Closure Dispensing | No |
Chemical Label | Ethyl Alcohol |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Color | White |
Unitary | Yes |
Certifications/Compliance | Meet OSHA requirements for workplace chemical container labeling. |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Unit Size | Case of 24 |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Through the Closure Dispensing | No |
Chemical Label | Ethyl Alcohol |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Color | White |
Unitary | Yes |
Certifications/Compliance | Meet OSHA requirements for workplace chemical container labeling. |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Unit Size | Pack of 4 |
Closure Material | HDPE |
Closure Type | Screw Cap |
Through the Closure Dispensing | No |
Chemical Label | Sodium Hypochlorite |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Color | White |
Unitary | Yes |
Certifications/Compliance | Meet OSHA requirements for workplace chemical container labeling. |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Unit Size | Case of 24 |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Chemical Label | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2436-1005PK also known as 2436-1005 | Pack of 2 | 1000 mL | Distilled Water | Price: 55.10 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2436-0503 | Case of 24 | 500 mL | Methanol | Price: 330.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2436-0503PK also known as 2436-0503 | Pack of 4 | 500 mL | Methanol | Price: 65.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2436-0504 | Case of 24 | 500 mL | Isopropanol | Price: 330.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2436-0504PK also known as 2436-0504 | Pack of 4 | 500 mL | Isopropanol | Price: 65.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2436-0505 | Case of 24 | 500 mL | Distilled Water | Price: 330.00 Online price: 264.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2436-0505PK also known as 2436-0505 | Pack of 4 | 500 mL | Distilled Water | Price: 65.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2436-0502 | Case of 24 | 500 mL | Ethyl Alcohol | Price: 330.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2436-0502PK also known as 2436-0502 | Pack of 4 | 500 mL | Ethyl Alcohol | Price: 65.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2436-0506 | Case of 24 | 500 mL | Sodium Hypochlorite | Price: 337.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|