Bacto Soytone is an enzymatic digest of soy that uses an animal origin (AO) (porcine) enzyme for digestion. The naturallyRead more
Have Questions?
Change view
Catalog Number
Quantity
243620
500 g
243610
10 kg
2 Options
Catalog number 243620
Price (MXN)
1,938.23
Each
Quantity:
500 g
Price (MXN)
1,938.23
Each
Bacto Soytone is an enzymatic digest of soy that uses an animal origin (AO) (porcine) enzyme for digestion. The naturally high concentrations of vitamins and carbohydrates found in soy make Bacto Soytone a nutritious and excellent carbohydrate source for those not concerned about the processing step involving an animal-based material. This supplement offers a unique nutritional profile as compared to other soy products, and so can be a valuable alternative to consider for supplementing both mammalian and microbial processes.
Specifications
Certifications/ComplianceLUO
For Use With (Application)Enzymatic Hydrolysate of Soybean meal
FormatBottle
Product LineBacto
Quantity500 g
Shelf Life1825 Days
SourceAnimal origin, porcine
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.