Potassium citrate tribasic monohydrate, 99%, extra pure
Potassium citrate tribasic monohydrate, 99%, extra pure
Potassium citrate tribasic monohydrate, 99%, extra pure
Thermo Scientific Chemicals

Potassium citrate tribasic monohydrate, 99%, extra pure

CAS: 6100-05-6 | C6H5K3O7·H2O | 324.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 244210050
Price (EUR)
189,00
Each
Quantity:
5 kg
Request bulk or custom format
Price (EUR)
189,00
Each
Chemical Identifiers
CAS6100-05-6
IUPAC Nametripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
Molecular FormulaC6H7K3O8
InChI KeyPJAHUDTUZRZBKM-UHFFFAOYSA-K
SMILESO.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Water4 to 6 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only