2-Hydroxynicotinic acid, 98%
2-Hydroxynicotinic acid, 98%
2-Hydroxynicotinic acid, 98%
Thermo Scientific Chemicals

2-Hydroxynicotinic acid, 98%

CAS: 609-71-2 | C6H5NO3 | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 247220250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS609-71-2
IUPAC Name2-oxo-1,2-dihydropyridine-3-carboxylic acid
Molecular FormulaC6H5NO3
InChI KeyUEYQJQVBUVAELZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CNC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to cream to light yellow
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Melting point256°C to 262°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only