(2S,3S)-(+)-2,3-Butanediol, 99%
(2S,3S)-(+)-2,3-Butanediol, 99%
(2S,3S)-(+)-2,3-Butanediol, 99%
Thermo Scientific Chemicals

(2S,3S)-(+)-2,3-Butanediol, 99%

CAS: 19132-06-0 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Catalog number 247730025
Price (USD)
-
Quantity:
2.5 g
Request bulk or custom format
Chemical Identifiers
CAS19132-06-0
IUPAC Name(2S,3S)-butane-2,3-diol
Molecular FormulaC4H10O2
InChI KeyOWBTYPJTUOEWEK-IMJSIDKUSA-N
SMILESC[C@H](O)[C@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4310 to 1.4340 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only