Methyl 6-methylnicotinate, 97%
Methyl 6-methylnicotinate, 97%
Methyl 6-methylnicotinate, 97%
Thermo Scientific Chemicals

Methyl 6-methylnicotinate, 97%

CAS: 5470-70-2 | C8H9NO2 | 151.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Packaging:
Glass bottle
Catalog number 248210100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5470-70-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
Infrared spectrumConforms
Appearance (Color)Brown
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only