1-Bromo-2,4-difluorobenzene, 98+%
1-Bromo-2,4-difluorobenzene, 98+%
1-Bromo-2,4-difluorobenzene, 98+%
Thermo Scientific Chemicals

1-Bromo-2,4-difluorobenzene, 98+%

CAS: 348-57-2 | C6H3BrF2 | 192.99 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
產品號碼 249360500
價格 (TWD)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS348-57-2
IUPAC Name1-bromo-2,4-difluorobenzene
Molecular FormulaC6H3BrF2
InChI KeyMGHBDQZXPCTTIH-UHFFFAOYSA-N
SMILESFC1=CC=C(Br)C(F)=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.0 %
Refractive index1.5040 to 1.5060 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only