Ethyl 4-bromobenzoate, 99%
Ethyl 4-bromobenzoate, 99%
Ethyl 4-bromobenzoate, 99%
Thermo Scientific Chemicals

Ethyl 4-bromobenzoate, 99%

CAS: 5798-75-4 | C9H9BrO2 | 229.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 249760500
Price (TWD)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5798-75-4
IUPAC Nameethyl 4-bromobenzoate
Molecular FormulaC9H9BrO2
InChI KeyXZIAFENWXIQIKR-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Refractive index1.5440 to 1.5470 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only