Thermo Scientific™
Superslip™ Cover Slips for Automated Coverslipping
 Related applications:
Anatomical Pathology
Minimize sticking with the uniquely developed Thermo Scientific™ Superslip™ Cover Slips, recommended for use in automatic coverslippers or for manual use in humid environments. All cover slips feature high-spectral transmission quality and a finely adaptive refractive index suited for microscopy. Each cover slip is individually selected for uniform quality in thickness, flatness, and size. These slides are made for use in anatomical pathology, hematology, microbiology, urinalysis, and research.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Virtually colorless appearance
  • Very low levels of inclusions, striae, bubbles, and streaks
  • High-spectral transmission
  • Finely adaptive refractive index suited for microscopes
  • Extremely flat surface
  • Packed 1 ounce per box, 10 ounces per case
Documents
Manuals & protocols