Odatrak Fibre-Optic to 4 to 20mA adapter
Thermo Fisher Scientific™

Odatrak Fibre-Optic to 4 to 20mA adapter

Odatrak Fibre-Optic to 4 to 20mA adapter allows an OdaLog L2 Logger, with fibre optic fitting, to be connected to a SCADA system - transmitting data as a 4-20mA signal.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo
25-0356Conversor de fibra óptica a 4-20 mA
12-0008Cable de fibra óptica, 5 m
12-0010Cable de fibra óptica, 25 m
Número de catálogo 25-0356
Precio (MXN)
-
Tipo:
Conversor de fibra óptica a 4-20 mA
Odatrak Fibre-Optic to 4 to 20mA adapter allows an OdaLog L2 Logger, with fibre optic fitting, to be connected to a SCADA system - transmitting data as a 4-20mA signal.

OdaTrak™ System

Key Features

  • Specifically designed for harsh wastewater environments: unique adapter fully sealed, proven & robust design
  • Quick & easy installation: standard M20 electrical thread, special machined housing for easy mounting of the OdaTrak™ optic cable
  • Programmed fault detection: OdaTrak detects loss of data from the OdaLog and will output a fault signal on the 4-20mA line alerting user to fault condition
  • Special fibre optic cable (ordered separately): safe to use in most commonly found hazardous environments. Two models are available, both with stainless steel connectors;
        1. standard coating for the fibre optic cable;
        2. extra protective sheath added over the cable for harsher wastewater environments.
  • Easy to transmit data: incorporates a 4-20mA adapter to transmit gas data using a fibre optic cable
  • Real time data: readily available when connected to a PLC network
  • Especificaciones
    Tipos de alarmaUnder-range, over-range, fault - as specific mA values
    Certificaciones/conformidadIP66
    Clasificación IPIP66
    IncluyeOdatrak Fibre-Optic to 4 to 20mA adapter
    TensiónLoop powered
    Peso (imperial)5.3 oz.
    Peso (métrico)150 g
    TipoConversor de fibra óptica a 4-20 mA
    Unit SizeEach