Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Thermo Scientific™

Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlate CompatibilitySterilityCondensation RingsCut-off Corners
250005All MicroplatesNon-sterileNoNo
250002All MicroplatesSterileNoNo
250003All MicroplatesNon-sterileNoNo
253623All MicroplatesNon-sterileYesNo
253624All MicroplatesSterileYesNo
26333996-well PlateNon-sterileYesLong Side
26412296-well PlateSterileYesLong Side
264611384-well PlateSterileNoLong Side
264612384-well PlateNon-sterileNoLong Side
264616384-well PlateSterileNoLong Side
26462396-well PlateSterileYesLong Side
Catalog number 250005
Price (MXN)
-
Plate Compatibility:
All Microplates
Sterility:
Non-sterile
Condensation Rings:
No
Cut-off Corners:
No
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
Specifications
Dimensions (L x W)5 x 3.35 in. (127 x 85 mm)
Dimensions (L x W x H)5 x 3.35 x 0.33 in. (127 x 85 x 8.34 mm)
Evaporation BarrierNo
Height (English)0.33 in.
Height (Metric)8.34 mm
Length (English)5 in.
Length (Metric)127 mm
No. per Case180
No. per Pack20
Product TypeUniversal Lid
Width (English)3.35 in.
Width (Metric)85 mm
AdhesiveNone
ColorClear
Condensation RingsNo
Cut-off CornersNo
MaterialPolystyrene
No. of Wells384/1536
Plate CompatibilityAll Microplates
Product LineNunc
SterilityNon-sterile
Temperature Range (Metric)-20°C to +80°C
Well ShapeAll well shapes
Unit SizeCase of 180