Hypersil GOLD™ Javelin™ Guard Columns
Hypersil GOLD™ Javelin™ Guard Columns
Offer convenient, easy to use protection for analytical HPLC columns
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호직경(미터법) 내부길이(미터법)입자 크기
25003-0121062.1 mm10 mm3 μm
25005-0121062.1 mm10 mm5 μm
25005-0140064 mm10 mm5 μm
카탈로그 번호 25003-012106
제품 가격(KRW)
-
직경(미터법) 내부:
2.1 mm
길이(미터법):
10 mm
입자 크기:
3 μm
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Hypersil GOLD HPLC columns provide excellent peak shape for all analyte types
  • Significant reduction in peak tailing while retaining C18 selectivity
  • Excellent resolution, efficiency and sensitvity
  • Confidence in the accuracy and quality of analytical data
사양
정지상C18
직경(미터법) 내부2.1 mm
길이(미터법)10 mm
입자 크기3 μm
제품라인Hypersil GOLD
Unit SizeEach